3-(Methacryloyloxy)propyltrimethoxysilane (MPS) acts as a functional comonomer, which is used to prepare polystyrene latex having silanol by emulsion polymerization. It is used in the preparation of polymers in association with other monomers like vinyl acetate, acrylic acid and methyl acrylic acid used in coating, adhesive and sealing agents, which provides excellent adhesion and durability. The composite materials made from unsaturated polyester by using MPS improve mechanical property.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(Methacryloyloxy)propyltrimethoxysilane (MPS) acts as a functional comonomer, which is used to prepare polystyrene latex having silanol by emulsion polymerization. It is used in the preparation of polymers in association with other monomers like vinyl acetate, acrylic acid and methyl acrylic acid used in coating, adhesive and sealing agents, which provides excellent adhesion and durability. The composite materials made from unsaturated polyester by using MPS improve mechanical property.
Solubility
Miscible with water.
Notes
Moisture sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only
Gharbharan, D.; Britsch, D.; Soto, G.; Weed, A. K.; Svec, F.; Zajickova, Z. Tuning preparation conditions towards optimized separation performance of thermally polymerized organo-silica monolithic columns in capillary liquid chromatography. J. Chromatogr. A 2015, 1408, 101-107.
Liu, Y.; Chen, R.; Yuan, D.; Liu, Z.; Meng, M.; Wang, Y.; Meng, J. H. X.; Liu, F, Hu, Z. Thermal-responsive ion-imprinted polymer based on magnetic mesoporous silica SBA-15 for selective removal of Sr(II) from aqueous solution. Colloid. Polym. Sci. 2015, 293 (1), 109-123.