Thermo Scientific Chemicals

4-Amino-1,2,4-triazole, 99%, Thermo Scientific Chemicals

Catalog number: A17716.22
100 g, Each
Thermo Scientific Chemicals

4-Amino-1,2,4-triazole, 99%, Thermo Scientific Chemicals

Catalog number: A17716.22
100 g, Each
Quantity
Catalog number: A17716.22
also known as A17716-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
584-13-4
IUPAC Name
4H-1,2,4-triazol-4-amine
Molecular Formula
C2H4N4
InChI Key
FMCUPJKTGNBGEC-UHFFFAOYSA-N
SMILES
NN1C=NN=C1
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Water Content (Karl Fischer Titration)
≤0.75%
Appearance (Color)
White
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder or fused solid

Description

4-Amino-1,2,4-triazole is used as an intermediate for the preparation of antifungal agent like fluconazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-1,2,4-triazole is used as an intermediate for the preparation of antifungal agent like fluconazole.

Solubility
Soluble in water and methanol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text