Thermo Scientific Chemicals

D-Fructose, 99%

Catalog number: A17718.0B
1000 g, Each
Thermo Scientific Chemicals

D-Fructose, 99%

Catalog number: A17718.0B
1000 g, Each
Quantity
Catalog number: A17718.0B
also known as A17718-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57-48-7
IUPAC Name
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
Molecular Formula
C6H12O6
InChI Key
BJHIKXHVCXFQLS-UYFOZJQFSA-N
SMILES
OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO
Appearance (Color)
White
Form
Crystals or crystalline powder
Water Content (Karl Fischer Titration)
<0.5% (non-U.S. specification)
Optical Rotation
-89.0° ± 3.0° (c=2 in water, non-U.S. specification)
Assay (unspecified)
≥98.5 to ≤101.5%

Description

Fructose is used commercially in foods and beverages. It increases starch viscosity more rapidly and achieves a higher final viscosity than sucrose. It has all the fiber squeezed out of it and so the fructose is easily absorbed into the bloodstream. High-fructose-corn syrup is used in frozen products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fructose is used commercially in foods and beverages. It increases starch viscosity more rapidly and achieves a higher final viscosity than sucrose. It has all the fiber squeezed out of it and so the fructose is easily absorbed into the bloodstream. High-fructose-corn syrup is used in frozen products.

Solubility
Soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text