DL-Glutamic acid monohydrate, 98+%, Thermo Scientific Chemicals
DL-Glutamic acid monohydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

DL-Glutamic acid monohydrate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number A17719.22
also known as A17719-22
Price (USD)/ Each
70.90
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
70.90
Add to cart
DL-Glutamic acid monohydrate, 98+%, Thermo Scientific Chemicals
Catalog numberA17719.22
Price (USD)/ Each
70.90
-
Add to cart
Chemical Identifiers
CAS19285-83-7
IUPAC Name2-aminopentanedioic acid hydrate
Molecular FormulaC5H11NO5
InChI KeyOZDAOHVKBFBBMZ-UHFFFAOYNA-N
SMILESO.NC(CCC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0% (dry wt. basis)
Optical Rotation0° ±0.9° (c=1 in water)
Appearance (Color)White
Water Content (Karl Fischer Titration)≤15.0% (theory 10.91%)
FormCrystalline powder or powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only