Cellulose, microcrystalline
Cellulose, microcrystalline
Thermo Scientific Chemicals

Cellulose, microcrystalline

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
500 g
10,000 g
Catalog number A17730.0E
also known as A17730-0E
Price (USD)/ Each
180.65
Online exclusive
200.00 
Save 19.35 (10%)
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
180.65
Online exclusive
200.00 
Save 19.35 (10%)
Add to cart
Cellulose, microcrystalline
Catalog numberA17730.0E
Price (USD)/ Each
180.65
Online exclusive
200.00 
Save 19.35 (10%)
-
Add to cart
Chemical Identifiers
CAS9004-34-6
IUPAC Name2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy}-6-(hydroxymethyl)-5-methoxyoxane-3,4-diol
Molecular FormulaC14H26O11
InChI KeyPTHCMJGKKRQCBF-UHFFFAOYNA-N
SMILESCOC1OC(CO)C(OC2OC(CO)C(OC)C(O)C2O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Bulk DensityBulk density: 0.25-0.36 g/cc
Appearance (Color)White to pale cream
Residue on ignition≤0.1%
Degree of PolymerizationDegree of polymerization ≤350
Loss on Drying≤7.0%
View more
Cellulose, microcrystalline is used as an emulsifier, bulking agent in food production, extender and anti-caking agent. It is also used in vitamin supplements or tablets. It acts as an alternative to carboxymethylcellulose. It finds application in partition chromatography. Further, it is used as a thickener and stabilizer. In addition to this, it is used as an excipient in the pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only