Thermo Scientific Chemicals

Cellulose, microcrystalline

Catalog number: A17730.36
500 g, Each
Thermo Scientific Chemicals

Cellulose, microcrystalline

Catalog number: A17730.36
500 g, Each
Quantity
Catalog number: A17730.36
also known as A17730-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9004-34-6
IUPAC Name
2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy}-6-(hydroxymethyl)-5-methoxyoxane-3,4-diol
Molecular Formula
C14H26O11
InChI Key
PTHCMJGKKRQCBF-UHFFFAOYNA-N
SMILES
COC1OC(CO)C(OC2OC(CO)C(OC)C(O)C2O)C(O)C1O
Bulk Density
Bulk density: 0.25-0.36 g/cc
Appearance (Color)
White to pale cream
Residue on ignition
≤0.1%
Degree of Polymerization
Degree of polymerization ≤350
Loss on Drying
≤7.0%

Description

Cellulose, microcrystalline is used as an emulsifier, bulking agent in food production, extender and anti-caking agent. It is also used in vitamin supplements or tablets. It acts as an alternative to carboxymethylcellulose. It finds application in partition chromatography. Further, it is used as a thickener and stabilizer. In addition to this, it is used as an excipient in the pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text