Thermo Scientific Chemicals

4,5-Dichloro-o-phenylenediamine, 98%, Thermo Scientific Chemicals

Catalog number: A17738.14
25 g, Each
Thermo Scientific Chemicals

4,5-Dichloro-o-phenylenediamine, 98%, Thermo Scientific Chemicals

Catalog number: A17738.14
25 g, Each
Quantity
Catalog number: A17738.14
also known as A17738-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5348-42-5
IUPAC Name
4,5-dichlorobenzene-1,2-diamine
Molecular Formula
C6H6Cl2N2
InChI Key
IWFHBRFJOHTIPU-UHFFFAOYSA-N
SMILES
NC1=CC(Cl)=C(Cl)C=C1N
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder or lumps
Appearance (Color)
Dark brown to brown or dark purple to purple

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol and Dimethyl sulfoxide. Insoluble in water.

Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text