Thermo Scientific Chemicals

1,3-Dihydroxynaphthalene, 98%, Thermo Scientific Chemicals

Catalog number: A17739.06
5 g, Each
Thermo Scientific Chemicals

1,3-Dihydroxynaphthalene, 98%, Thermo Scientific Chemicals

Catalog number: A17739.06
5 g, Each
Quantity
Catalog number: A17739.06
also known as A17739-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
132-86-5
IUPAC Name
naphthalene-1,3-diol
Molecular Formula
C10H8O2
InChI Key
XOOMNEFVDUTJPP-UHFFFAOYSA-N
SMILES
OC1=CC(O)=C2C=CC=CC2=C1
Appearance (Color)
Cream to brown
Assay (Silylated GC)
≥97.5%
Assay (HPLC)
≥97.5%
Melting Point (clear melt)
121.0-128.0?C
Form
Powder

Description

It is used in the naphthoresorcinol test for uronic acids of Tollens and Rorive, a blue color can be extracted with ethanol after heating of uronic acids with naphthoresorcinol in hydrochloric acid. It is also employed as a reagent for various medical tests. It can also be used as a spot test reagent for glycerate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the naphthoresorcinol test for uronic acids of Tollens and Rorive, a blue color can be extracted with ethanol after heating of uronic acids with naphthoresorcinol in hydrochloric acid. It is also employed as a reagent for various medical tests. It can also be used as a spot test reagent for glycerate.

Solubility
Soluble in ethanol (50 mg/ml), and water.

Notes
Light Sensitive. Store away from oxidizing agents and bases. Keep the container tightly closed in a cool, dry and well ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text