m-Tolylacetic acid, 99%, Thermo Scientific Chemicals
m-Tolylacetic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

m-Tolylacetic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number A17743.18
also known as A17743-18
Price (USD)/ Each
137.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
137.00
Add to cart
m-Tolylacetic acid, 99%, Thermo Scientific Chemicals
Catalog numberA17743.18
Price (USD)/ Each
137.00
-
Add to cart
Chemical Identifiers
CAS621-36-3
IUPAC Name2-(3-methylphenyl)acetic acid
Molecular FormulaC9H10O2
InChI KeyGJMPSRSMBJLKKB-UHFFFAOYSA-N
SMILESCC1=CC=CC(CC(O)=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Melting Point (clear melt)60.0-67.0?C
FormCrystals or powder or crystalline powder or fused solid
Identification (FTIR)Conforms
Appearance (Color)White to cream
View more
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Amin A. Nomeir; and Mohamed B. Abou-Donia. High-performance liquid chromatographic analysis on radial compression column of the neurotoxic tri-o-cresyl phosphate and metabolites. Analytical Biochemistry, 1983 135 (2), 296-303.
  2. K Bernhard and R Viret. 223. Further studies on the metabolic behavior of some aromatic and hydroaromatic mono- and dicarboxylic acids. Hoppe-Seyler's Zeitschrift für physiologische Chemie, 1958, 310(1-2), 37-43 .