2,2':6',2″-Terpyridine, 97%, Thermo Scientific Chemicals
2,2':6',2″-Terpyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2':6',2″-Terpyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
0.25 g
5 g
Catalog number A17750.03
also known as A17750-03
Price (USD)/ Each
135.65
Online exclusive
151.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
135.65
Online exclusive
151.00 
Save 15.35 (10%)
Add to cart
2,2':6',2″-Terpyridine, 97%, Thermo Scientific Chemicals
Catalog numberA17750.03
Price (USD)/ Each
135.65
Online exclusive
151.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1148-79-4
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)85.5-94.5?C
FormPowder
Assay (GC)≥96.0%
Appearance (Color)Pale cream or pale yellow to orange to brown
2,2';6',2'-terpyridine is widely utilized in the field of supramolecular chemistry, which is used to manufacture the racks, ladders and grids, helicates, catenanes and dendrimers. It plays an important role as a ligand in coordination chemistry. Its complexes are employed in the oxidation of alcohols, the carbonylation of aromatic compounds and as oxygen-binding molecules. Functionalized terpyridine ligands were used in semiconductors and solar panels. It forms chealate complexes with europium(III) and terbium(III), which is used in protein labelling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′;6′,2″-terpyridine is widely utilized in the field of supramolecular chemistry, which is used to manufacture the racks, ladders and grids, helicates, catenanes and dendrimers. It plays an important role as a ligand in coordination chemistry. Its complexes are employed in the oxidation of alcohols, the carbonylation of aromatic compounds and as oxygen-binding molecules. Functionalized terpyridine ligands were used in semiconductors and solar panels. It forms chealate complexes with europium(III) and terbium(III), which is used in protein labelling.

Solubility
Soluble in most organic solvents.

Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Schönle, J.; Constable, E. C.; Housecroft, C. E.; Prescimone, A.; Zampese, J. E. Homoleptic and heteroleptic complexes of chromium(III) containing 4'-diphenylamino-2,2':6',2″-terpyridine ligands. Polyhedron 2015, 89 182-188.
  2. Harper, B. W. J.; Aldrich-Wright, J. R. The synthesis, characterisation and cytotoxicity of bisintercalating (2,2':6',2″-terpyridine)platinum(II) complexes. Dalton Trans. 2015, 44 (1), 87-96.