Thermo Scientific Chemicals

2,2':6',2''-Terpyridine, 97%, Thermo Scientific Chemicals

Catalog number: A17750.06
5 g, Each
Thermo Scientific Chemicals

2,2':6',2''-Terpyridine, 97%, Thermo Scientific Chemicals

Catalog number: A17750.06
5 g, Each
Quantity
Catalog number: A17750.06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1148-79-4
Melting Point (clear melt)
85.5-94.5?C
Form
Powder
Assay (GC)
≥96.0%
Appearance (Color)
Pale cream or pale yellow to orange to brown

Description

2,2';6',2'-terpyridine is widely utilized in the field of supramolecular chemistry, which is used to manufacture the racks, ladders and grids, helicates, catenanes and dendrimers. It plays an important role as a ligand in coordination chemistry. Its complexes are employed in the oxidation of alcohols, the carbonylation of aromatic compounds and as oxygen-binding molecules. Functionalized terpyridine ligands were used in semiconductors and solar panels. It forms chealate complexes with europium(III) and terbium(III), which is used in protein labelling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′;6′,2″-terpyridine is widely utilized in the field of supramolecular chemistry, which is used to manufacture the racks, ladders and grids, helicates, catenanes and dendrimers. It plays an important role as a ligand in coordination chemistry. Its complexes are employed in the oxidation of alcohols, the carbonylation of aromatic compounds and as oxygen-binding molecules. Functionalized terpyridine ligands were used in semiconductors and solar panels. It forms chealate complexes with europium(III) and terbium(III), which is used in protein labelling.

Solubility
Soluble in most organic solvents.

Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text