Thermo Scientific Chemicals

TAPS, 99%, Thermo Scientific Chemicals

Catalog number: A17754.14
25 g, Each
Thermo Scientific Chemicals

TAPS, 99%, Thermo Scientific Chemicals

Catalog number: A17754.14
25 g, Each
Quantity
Catalog number: A17754.14
also known as A17754-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
29915-38-6
IUPAC Name
3-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}propane-1-sulfonic acid
Molecular Formula
C7H17NO6S
InChI Key
YNLCVAQJIKOXER-UHFFFAOYSA-N
SMILES
OCC(CO)(CO)NCCCS(O)(=O)=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

A zwitterionic Good's Buffer

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text