Thermo Scientific Chemicals

1-Pyrenebutyric acid, 97%, Thermo Scientific Chemicals

Catalog number: A17760.03
1 g, Each
Thermo Scientific Chemicals

1-Pyrenebutyric acid, 97%, Thermo Scientific Chemicals

Catalog number: A17760.03
1 g, Each
Quantity
Catalog number: A17760.03
also known as A17760-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3443-45-6
Appearance (Color)
Pale yellow to yellow to brown
Form
Powder
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (HPLC)
≥96.0%
Identification (FTIR)
Conforms

Description

1-Pyrenebutyric acid is a fluorescent probe used to study proteins, lipids, nucleic acids and other biological systems. It acts as a UV excitable fluorophore that has concentration-dependent fluorescence. It is also employed as a probe for bovine serum. 1-pyrene butyric acid modified nitrogen-doped carbon nanotubes (PBA/NCNTs) nanocomposites can be used as a biosensor to detect glucose in human serum sample.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pyrenebutyric acid is a fluorescent probe used to study proteins, lipids, nucleic acids and other biological systems. It acts as a UV excitable fluorophore that has concentration-dependent fluorescence. It is also employed as a probe for bovine serum. 1-pyrene butyric acid modified nitrogen-doped carbon nanotubes (PBA/NCNTs) nanocomposites can be used as a biosensor to detect glucose in human serum sample.

Solubility
Partially soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text