N-Methylcaprolactam was used to study the inhibition mechanism of poly-[N-vinylcaprolactam] (PVCAP) and its related compounds against hydrate formation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Methylcaprolactam was used to study the inhibition mechanism of poly-[N-vinylcaprolactam] (PVCAP) and its related compounds against hydrate formation.
Solubility
Fully miscible with water.
Notes
Hygroscopic, store away from moisture. Store in a tightly closed container in a cool, dry, well-ventilated area. Store under dry inert gas.
RUO – Research Use Only
Michika Ohtake et. al .Clustering structure of aqueous solution of kinetic inhibitor of gas hydrates. Journal of Physical Chemistry B, 2005,109(35), 16879-16885.
Richard E. Benson; Theodore L. Cairns. Chemical Reactions of Caprolactam. J. Am. Chem. Soc, 1948,70(6), 2115-2118.