Thermo Scientific Chemicals

Benzo[h]quinoline, 98%, Thermo Scientific Chemicals

Catalog number: A17794.06
5 g, Each
Thermo Scientific Chemicals

Benzo[h]quinoline, 98%, Thermo Scientific Chemicals

Catalog number: A17794.06
5 g, Each
Quantity
Catalog number: A17794.06
also known as A17794-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
230-27-3
IUPAC Name
benzo[h]quinoline
Molecular Formula
C13H9N
InChI Key
WZJYKHNJTSNBHV-UHFFFAOYSA-N
SMILES
C1=CC=C2C(C=CC3=CC=CN=C23)=C1
Appearance (Color)
White to yellow to pale brown
Assay (GC)
≥97.5%
Melting Point (clear melt)
45.0-53.0°C
Form
Crystals or powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text