Thermo Scientific Chemicals

D-(+)-Arabitol, 99%

Catalog number: A17801.14
25 g, Each
Thermo Scientific Chemicals

D-(+)-Arabitol, 99%

Catalog number: A17801.14
25 g, Each
Quantity
Catalog number: A17801.14
also known as A17801-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
488-82-4
IUPAC Name
pentane-1,2,3,4,5-pentol
Molecular Formula
C5H12O5
InChI Key
HEBKCHPVOIAQTA-UHFFFAOYNA-N
SMILES
OCC(O)C(O)C(O)CO
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥98.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
100.0-107.0?C
Optical Rotation
9.5 ± 0.5? (C=5 in 4% borax solution)

Description

It can be employed as a sugar in the production of xylitol via biotransformation process. It is a substrate used to identify, differentiate and characterize enzyme such as the gluconobacter oxydans dehydrogenase(s), Gox2181, hyperthermophilic D-arabitol dehydrogenase from Thermotoga maritime and NAD-dependent D-arabitoldehydrogenase from acetic acid bacterium, Acetobacter suboxydans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be employed as a sugar in the production of xylitol via biotransformation process. It is a substrate used to identify, differentiate and characterize enzyme such as the gluconobacter oxydans dehydrogenase(s), Gox2181, hyperthermophilic D-arabitol dehydrogenase from Thermotoga maritime and NAD-dependent D-arabitoldehydrogenase from acetic acid bacterium, Acetobacter suboxydans.

Solubility
Soluble in water (50 mg/ml).

Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry and well ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text