Thermo Scientific Chemicals

Methanesulfonic acid, 70% aq. soln., Thermo Scientific Chemicals

Catalog number: A17803.AE
100 mL, Each
Thermo Scientific Chemicals

Methanesulfonic acid, 70% aq. soln., Thermo Scientific Chemicals

Catalog number: A17803.AE
100 mL, Each
Quantity
Catalog number: A17803.AE
also known as A17803-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-75-2
IUPAC Name
methanesulfonic acid
Molecular Formula
CH4O3S
InChI Key
AFVFQIVMOAPDHO-UHFFFAOYSA-N
SMILES
CS(O)(=O)=O
Appearance (Color)
Clear colorless
Form
Liquid
Assay (Aqueous acid-base Titration)
≥68.5 to ≤71.5%

Description

Methane sulfonic acid 70% solution is used virtually in all electronic applications involving tin or tin-alloy plating from non fluoborate systems. It replaces phosphoric acid in detergent formulations, thereby helping to avoid environmental phosphate pollution. It is used in the preparation of polyaniline/graphene nanocomposites, which enhances its thermal and electrical properties. It is utilized for electropolishing of aluminum as well as in electroplating of noble metals like gold, platinum, palladium and their alloys.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methane sulfonic acid 70% solution is used virtually in all electronic applications involving tin or tin-alloy plating from non fluoborate systems. It replaces phosphoric acid in detergent formulations, thereby helping to avoid environmental phosphate pollution. It is used in the preparation of polyaniline/graphene nanocomposites, which enhances its thermal and electrical properties. It is utilized for electropolishing of aluminum as well as in electroplating of noble metals like gold, platinum, palladium and their alloys.

Solubility
Miscible with water. Slightly miscible with benzene and toluene. Immiscible with paraffins.

Notes
Light and heat sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with amines, strong reducing agents, ethyl vinyl ether, hydrofluoric acid, strong oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text