Thermo Scientific Chemicals

Sodium formate, 98%, Thermo Scientific Chemicals

Catalog number: A17813.30
250 g, Each
Thermo Scientific Chemicals

Sodium formate, 98%, Thermo Scientific Chemicals

Catalog number: A17813.30
250 g, Each
Quantity
Catalog number: A17813.30
also known as A17813-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
141-53-7
IUPAC Name
sodium formate
Molecular Formula
CHNaO2
InChI Key
HLBBKKJFGFRGMU-UHFFFAOYSA-M
SMILES
[Na+].[O-]C=O
Appearance (Color)
White or pale cream
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤1.0%

Description

Sodium formate is used in analytical chemistry to precipitate noble metals. It is used for tanning leather, for dyeing and printing fabrics. It acts as an electroplating agent, an acidulant in the textile industry, a reducing agent, a mordant, a complexing agent. Further, it is used in photographic fixing baths. In addition, it is used as a buffering agent for strong mineral acids and food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium formate is used in analytical chemistry to precipitate noble metals. It is used for tanning leather, for dyeing and printing fabrics. It acts as an electroplating agent, an acidulant in the textile industry, a reducing agent, a mordant, a complexing agent. Further, it is used in photographic fixing baths. In addition, it is used as a buffering agent for strong mineral acids and food additive.

Solubility
Soluble in water, alcohol, glycerol and formic acid. Insoluble in ether.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong acids. Moisture sensitive
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text