Thermo Scientific Chemicals

(1S)-(-)-beta-Pinene, 99%, Thermo Scientific Chemicals

Catalog number: A17818.AE
100 mL, Each
Thermo Scientific Chemicals

(1S)-(-)-beta-Pinene, 99%, Thermo Scientific Chemicals

Catalog number: A17818.AE
100 mL, Each
Quantity
Catalog number: A17818.AE
also known as A17818-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18172-67-3
IUPAC Name
(1S,5S)-6,6-dimethyl-2-methylidenebicyclo[3.1.1]heptane
Molecular Formula
C10H16
InChI Key
WTARULDDTDQWMU-IUCAKERBSA-N
SMILES
CC1(C)[C@@H]2C[C@H]1C(=C)CC2
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.4775-1.4805 @ 20?C
Appearance (Color)
Clear colorless
Optical Rotation
-20.5 ± 2.0? (neat)

Description

It is a useful biochemical for proteomics research. It is also employed in the preparation of natural products, such as grandisol and robustadial. It is also applied as perfuming agent in cosmetics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful biochemical for proteomics research. It is also employed in the preparation of natural products, such as grandisol and robustadial. It is also applied as perfuming agent in cosmetics industry.

Solubility
Not miscible or difficult to mix in water, 7.061 mg/L @ 25°C (est). Soluble in alcohol, chloroform and ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text