Thermo Scientific Chemicals

Cyclooctanol, 97%, Thermo Scientific Chemicals

Catalog number: A17825.14
25 g, Each
Thermo Scientific Chemicals

Cyclooctanol, 97%, Thermo Scientific Chemicals

Catalog number: A17825.14
25 g, Each
Quantity
Catalog number: A17825.14
also known as A17825-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
696-71-9
IUPAC Name
cyclooctanol
Molecular Formula
C8H16O
InChI Key
FHADSMKORVFYOS-UHFFFAOYSA-N
SMILES
OC1CCCCCCC1
Refractive Index
1.4835-1.4885 @ 20°C
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥96.0%

Description

Cyclooctanol is a key starting material used for the preparation of cylooctanone by using a chromic acid reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclooctanol is a key starting material used for the preparation of cylooctanone by using a chromic acid reagent.

Solubility
Miscible with acetone.

Notes
Contains 75+% cyclopentane; remainder consists of C{5} and C{6} hydrocarbon.Incompatible with strong oxidizing agents, acid chloride, and acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text