Thermo Scientific Chemicals

N,N-Diethyl-p-phenylenediamine sulfate, 97%, Thermo Scientific Chemicals

Catalog number: A17832.18
50 g, Each
Thermo Scientific Chemicals

N,N-Diethyl-p-phenylenediamine sulfate, 97%, Thermo Scientific Chemicals

Catalog number: A17832.18
50 g, Each
Quantity
Catalog number: A17832.18
also known as A17832-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6283-63-2
IUPAC Name
N1,N1-diethylbenzene-1,4-diamine; sulfuric acid
Molecular Formula
C10H18N2O4S
InChI Key
AYLDJQABCMPYEN-UHFFFAOYSA-N
SMILES
OS(O)(=O)=O.CCN(CC)C1=CC=C(N)C=C1
Appearance (Color)
White to pale brown
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Melting Point (clear melt)
182-189?C
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%

Description

N,N-Diethyl-p-phenylenediamine sulfate is used for the spectrophotometric determination of drugs such as salbutamol sulphate, ritodrine hydrochloride and isoxsuprine hydrochloride containing phenolic group. It is also used in the analysis of drugs like dapsone hydrochloride, sulfamethoxazole and sulfadiazine containing aromatic amino group. Further, it is used as a color developer and film developer in photography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diethyl-p-phenylenediamine sulfate is used for the spectrophotometric determination of drugs such as salbutamol sulphate, ritodrine hydrochloride and isoxsuprine hydrochloride containing phenolic group. It is also used in the analysis of drugs like dapsone hydrochloride, sulfamethoxazole and sulfadiazine containing aromatic amino group. Further, it is used as a color developer and film developer in photography.

Solubility
Soluble in water and alcohol.

Notes
Light sensitive and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text