Thermo Scientific Chemicals

o-Tolidine, 98%, Thermo Scientific Chemicals

Catalog number: A17837.14
25 g, Each
Thermo Scientific Chemicals

o-Tolidine, 98%, Thermo Scientific Chemicals

Catalog number: A17837.14
25 g, Each
Quantity
Catalog number: A17837.14
also known as A17837-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
119-93-7
IUPAC Name
3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diamine
Molecular Formula
C14H16N2
InChI Key
NUIURNJTPRWVAP-UHFFFAOYSA-N
SMILES
CC1=CC(=CC=C1N)C1=CC=C(N)C(C)=C1
Water Content
<5.0%
Appearance (Color)
White to cream to grey to pale brown
Form
Powder or chunks/lumps
Assay (GC)
≥97.5
Comment
Contains trace insolubles, solution in DMF is hazy

Description

O-Tolidine is widely used in dye production. It is particularly used in textile, leather and paper industries. It can be used as an indicator in analytical, clinical and forensic chemistry. It is utilized for the determination of gold and chlorine level in swimming pool water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-Tolidine is widely used in dye production. It is particularly used in textile, leather and paper industries. It can be used as an indicator in analytical, clinical and forensic chemistry. It is utilized for the determination of gold and chlorine level in swimming pool water.

Solubility
Slightly soluble in water.

Notes
Light and air sensitive. Incompatible with strong oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text