Thermo Scientific Chemicals

Gibberellic acid, 90+%, Thermo Scientific Chemicals

Catalog number: A17843.06
5 g, Each
Thermo Scientific Chemicals

Gibberellic acid, 90+%, Thermo Scientific Chemicals

Catalog number: A17843.06
5 g, Each
Quantity
Catalog number: A17843.06
also known as A17843-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77-06-5
IUPAC Name
(1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadec-13-ene-9-carboxylic acid
Molecular Formula
C19H22O6
InChI Key
IXORZMNAPKEEDV-QTWFBFKQSA-N
SMILES
CC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O
Form
Powder
Optical Rotation
78.0 ± 2.5? (C=2 in methanol)
Assay (Aqueous acid-base Titration)
≥90.0 to ≤110.0%
Identification (FTIR)
Conforms
Appearance (Color)
White to cream to pale brown

Description

It is a plant growth regulating hormone that promotes plant cell growth and elongation. They promote rapid stem and root growth, induce mitotic division and initiate (break dormancy) and increase seed germination rates. They are also involved in processes such as gravitropism, tensioning and floral display. Gibberellin A3 (G1025, G7645) and A4 (G7276) may be used as supplements to plant growth media such as Murashige and Skoog media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a plant growth regulating hormone that promotes plant cell growth and elongation. They promote rapid stem and root growth, induce mitotic division and initiate (break dormancy) and increase seed germination rates. They are also involved in processes such as gravitropism, tensioning and floral display. Gibberellin A3 (G1025, G7645) and A4 (G7276) may be used as supplements to plant growth media such as Murashige and Skoog media.

Solubility
Soluble in water 20°C 5g/L. Soluble in Ethanol: 200 mg/4ml.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text