Thermo Scientific Chemicals

Pyridoxal hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A17855.06
5 g, Each
Thermo Scientific Chemicals

Pyridoxal hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A17855.06
5 g, Each
Quantity
Catalog number: A17855.06
also known as A17855-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
65-22-5
IUPAC Name
hydrogen 3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde chloride
Molecular Formula
C8H10ClNO3
InChI Key
FCHXJFJNDJXENQ-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CC1=NC=C(CO)C(C=O)=C1O
Appearance (Color)
White to cream
Form
powder
Assay (Titration ex Chloride)
> 98.5%

Description

Pyridoxal hydrochloride is used in labeling of amino acids for their detection. It finds application in neurotransmitters, sphingolipids and aminolevulinic acid. It is also used as a nutritional agent. It is also applied in research studies like biochemical, drugs and in media supplements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridoxal hydrochloride is used in labeling of amino acids for their detection. It finds application in neurotransmitters, sphingolipids and aminolevulinic acid. It is also used as a nutritional agent. It is also applied in research studies like biochemical, drugs and in media supplements.

Solubility
Soluble in water and ethanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong acids, strong reducing agents and strong bases. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text