Pyridoxal hydrochloride is used in labeling of amino acids for their detection. It finds application in neurotransmitters, sphingolipids and aminolevulinic acid. It is also used as a nutritional agent. It is also applied in research studies like biochemical, drugs and in media supplements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pyridoxal hydrochloride is used in labeling of amino acids for their detection. It finds application in neurotransmitters, sphingolipids and aminolevulinic acid. It is also used as a nutritional agent. It is also applied in research studies like biochemical, drugs and in media supplements.
Solubility
Soluble in water and ethanol.
Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong acids, strong reducing agents and strong bases. Store in a cool place.
RUO – Research Use Only
Kanchanadevi, A.; Ramesh, R.; Semeril, D. Synthesis of Ru(II) pyridoxal thiosemicarbazone complex and its catalytic application to one-pot conversion of aldehydes to primary amides. Inorg. Chem. Commun. 2015, 56, 116-119.
Hassan, B.; Shireen, A.; Muraleedharan, K.; Mujeeb, V. M. A. Virtual screening of molecular properties of chitosan and derivatives in search for druggable molecules. Int. J. Biol. Macromol. 2015, 74, 392-396.