4-Bromo-2-chloroaniline, 98+%, Thermo Scientific Chemicals
4-Bromo-2-chloroaniline, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromo-2-chloroaniline, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A17859.09
also known as A17859-09
Price (USD)/ Each
46.30
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
46.30
Add to cart
4-Bromo-2-chloroaniline, 98+%, Thermo Scientific Chemicals
Catalog numberA17859.09
Price (USD)/ Each
46.30
-
Add to cart
Chemical Identifiers
CAS38762-41-3
IUPAC Name3-bromo-5-chloroaniline
Molecular FormulaC6H5BrClN
InChI KeyYYBRLVWWPRAQDX-UHFFFAOYSA-N
SMILESNC1=CC(Cl)=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to brown
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Melting Point (clear melt)67.0-73.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only