Thermo Scientific Chemicals

Tetramethylammonium fluoride tetrahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A17864.14
25 g, Each
Thermo Scientific Chemicals

Tetramethylammonium fluoride tetrahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A17864.14
25 g, Each
Quantity
Catalog number: A17864.14
also known as A17864-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17787-40-5
IUPAC Name
tetramethylazanium tetrahydrate fluoride
Molecular Formula
C4H20FNO4
InChI Key
HQFTZNVQVRRDLN-UHFFFAOYSA-M
SMILES
O.O.O.O.[F-].C[N+](C)(C)C
Appearance (Color)
Colorless to white to pale cream
Form
Crystals or powder or crystalline powder or fused solid
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

It is employed as a reactant for homocoupling of aryl halides via a palladium catalyzed mechanism, in cross-coupling reactions with alkenylsilanols and in fluorination of dichloronitrobenzene. It is also used as a reversible absorbing agent for large quantities of hydrogen sulfide in the liquid state.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a reactant for homocoupling of aryl halides via a palladium catalyzed mechanism, in cross-coupling reactions with alkenylsilanols and in fluorination of dichloronitrobenzene. It is also used as a reversible absorbing agent for large quantities of hydrogen sulfide in the liquid state.

Solubility
Soluble in water.

Notes
Hygroscopic. Keep away from oxidizing agents, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. This product may become liquid during storage but it does not affect the quality of the product.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text