It is employed as a reactant for homocoupling of aryl halides via a palladium catalyzed mechanism, in cross-coupling reactions with alkenylsilanols and in fluorination of dichloronitrobenzene. It is also used as a reversible absorbing agent for large quantities of hydrogen sulfide in the liquid state.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a reactant for homocoupling of aryl halides via a palladium catalyzed mechanism, in cross-coupling reactions with alkenylsilanols and in fluorination of dichloronitrobenzene. It is also used as a reversible absorbing agent for large quantities of hydrogen sulfide in the liquid state.
Solubility
Soluble in water.
Notes
Hygroscopic. Keep away from oxidizing agents, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. This product may become liquid during storage but it does not affect the quality of the product.
RUO – Research Use Only
General References:
- Scott E Denmark.; Ramzi F Sweis.Cross-coupling reactions of alkenylsilanols with fluoroalkylsulfonates.Org Lett.,2002,4(21), 3771-3774.
- W. Michael Seganish.; Molly E. Mowery.; Shaundra Riggleman.; Philip DeShong. Palladium-catalyzed homocoupling of aryl halides in the presence of fluoride.Tetrahedron.;2005,61(8), 2117-2121.
- R. Quinn.; J. B. Appleby & G. P. Pez. Hydrogen sulfide separation from gas streams using salt hydrate chemical absorbents and immobilized liquid membranes.Sep. Sci. Technol.;2002,37(3), 627-638.