Thermo Scientific Chemicals

Thiodiglycolic acid, 98%, Thermo Scientific Chemicals

Catalog number: A17880.36
500 g, Each
Thermo Scientific Chemicals

Thiodiglycolic acid, 98%, Thermo Scientific Chemicals

Catalog number: A17880.36
500 g, Each
Quantity
Catalog number: A17880.36
also known as A17880-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123-93-3
IUPAC Name
2-[(carboxymethyl)sulfanyl]acetic acid
Molecular Formula
C4H6O4S
InChI Key
UVZICZIVKIMRNE-UHFFFAOYSA-N
SMILES
OC(=O)CSCC(O)=O
Appearance (Color)
White to pale cream
Identification (FTIR)
Conforms
Melting Point (clear melt)
129.0-135.0?C
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥97.5%

Description

It finds it applications in the production of [1,4]oxathiane-2,6-dione by heating. It is employed as an anti-oxidant and reducing agent in cosmetics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it applications in the production of [1,4]oxathiane-2,6-dione by heating. It is employed as an anti-oxidant and reducing agent in cosmetics industry.

Solubility
Solubility in water is almost transparent. (water, 4.00E+05 mg/L @ 25°C (exp); 1e+006 mg/L @ 25°C (est).

Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text