Thermo Scientific Chemicals

Trimethyl-p-tolylsilane, 95%, Thermo Scientific Chemicals

Catalog number: A17884.06
5 g, Each
Thermo Scientific Chemicals

Trimethyl-p-tolylsilane, 95%, Thermo Scientific Chemicals

Catalog number: A17884.06
5 g, Each
Quantity
Catalog number: A17884.06
also known as A17884-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3728-43-6
IUPAC Name
trimethyl(4-methylphenyl)silane
Molecular Formula
C10H16Si
InChI Key
QGHURGPPCGMAMZ-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)[Si](C)(C)C
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%
Refractive Index
1.4885-1.4935 @ 20?C
Form
Liquid

Description

It is employed in the production of trimethyl-(4-trimethylsilanyl-benzyl)-silane by reacting with chloro-trimethyl-silane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the production of trimethyl-(4-trimethylsilanyl-benzyl)-silane by reacting with chloro-trimethyl-silane.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect against electrostatic charges. Keep ignition sources away.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text