Thermo Scientific Chemicals

D-(-)-Ribose, 98%

Catalog number: A17894.30
250 g, Each
Thermo Scientific Chemicals

D-(-)-Ribose, 98%

Catalog number: A17894.30
250 g, Each
Quantity
Catalog number: A17894.30
also known as A17894-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50-69-1
IUPAC Name
(3R,4S,5R)-5-(hydroxymethyl)oxolane-2,3,4-triol
Molecular Formula
C5H10O5
InChI Key
HMFHBZSHGGEWLO-ZHZWXSSZNA-N
SMILES
OC[C@H]1OC(O)[C@H](O)[C@@H]1O
Appearance (Color)
White to cream to yellow to pale brown
Form
Powder or crystalline powder
Assay (Silylated GC)
≥97.5%
Identification (FTIR)
Conforms
Optical Rotation
-18° to -22° (c=10 in water, NH4OH)

Description

D-(-)-Ribose is mainly used as a very basic source of metabolism of life. It is also used as a precursor for virazole, adenosine, thymidine, cytidine, fluoro-thymidine, 2-methyl adenosine, pyrromonazole toxina and adenosine methionine. It is further used in the synthesis of vitamin B2 (riboflavin), tetra-O-acetyl-ribose and nucleoside. It finds application as food additives and as supplement in cell culture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(-)-Ribose is mainly used as a very basic source of metabolism of life. It is also used as a precursor for virazole, adenosine, thymidine, cytidine, fluoro-thymidine, 2-methyl adenosine, pyrromonazole toxina and adenosine methionine. It is further used in the synthesis of vitamin B2 (riboflavin), tetra-O-acetyl-ribose and nucleoside. It finds application as food additives and as supplement in cell culture.

Solubility
Soluble in water. Insoluble in ether.

Notes
Hygroscopic. Incompatible with strong oxidizing agents. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text