Thermo Scientific Chemicals

Phenylsilane, 97%, Thermo Scientific Chemicals

Catalog number: A17901.06
5 g, Each
Thermo Scientific Chemicals

Phenylsilane, 97%, Thermo Scientific Chemicals

Catalog number: A17901.06
5 g, Each
Quantity
Catalog number: A17901.06
also known as A17901-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
694-53-1
IUPAC Name
phenylsilyl
Molecular Formula
C6H7Si
InChI Key
JAZKZOWBOMBYIX-UHFFFAOYSA-N
SMILES
[SiH2]C1=CC=CC=C1
Refractive Index
1.5085-1.5135 @ 20°C (U.K. specification)
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥96.0% (U.K. specification)
Comment
Purchased in U.K. and U.S.

Description

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Contact with water releases a flammable alcohol. Soluble in water (Contact with water releases flammable gases), and most common organic solvents.

Notes
Moisture Sensitive. Store away from water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect against electrostatic charges. Keep ignition sources away. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text