Thermo Scientific Chemicals

Wright's Stain, Thermo Scientific Chemicals

Catalog number: A17903.22
100 g, Each
Thermo Scientific Chemicals

Wright's Stain, Thermo Scientific Chemicals

Catalog number: A17903.22
100 g, Each
Quantity
Catalog number: A17903.22
also known as A17903-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
68988-92-1
IUPAC Name
2,4,5,7-tetrabromo-9-(2-carboxyphenyl)-3,6-dihydroxy-10λ⁴-xanthen-10-ylium; 7-(dimethylamino)-N,N-dimethyl-3H-phenothiazin-3-iminium
Molecular Formula
C36H27Br4N3O5S
InChI Key
AXIKDPDWFVPGOD-UHFFFAOYSA-O
SMILES
CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C.OC(=O)C1=CC=CC=C1C1=C2C=C(Br)C(O)=C(Br)C2=[O+]C2=C(Br)C(O)=C(Br)C=C12
lambda-max (UV-Vis)
641-649nm (in methanol)
Appearance (Color)
Dark green
Form
Powder
Loss on Drying
≤10% (105?C/constant wt.)
Specific Absorbance (E 1%/1cm)
≥1100 at Lambda max

Description

Useful stain for blood and for bone marrow films

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text