Thermo Scientific Chemicals

Sodium thiosulfate pentahydrate, 99+%, Thermo Scientific Chemicals

Catalog number: A17914.0C
10000 g, Each
Thermo Scientific Chemicals

Sodium thiosulfate pentahydrate, 99+%, Thermo Scientific Chemicals

Catalog number: A17914.0C
10000 g, Each
Quantity
Catalog number: A17914.0C
also known as A17914-0C
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10102-17-7
IUPAC Name
disodium pentahydrate sulfanidesulfonate
Molecular Formula
H10Na2O8S2
InChI Key
PODWXQQNRWNDGD-UHFFFAOYSA-L
SMILES
O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
Form
Crystals or powder or crystalline powder or pellets
Assay (Redox Titration)
≥99.0 to ≤101.0%
Identification (FTIR)
Conforms
Appearance (Color)
Colorless to white

Description

Sodium thiosulfate pentahydrate is used as an analytical reagent in laboratories. It is used in photographic processing and in gold extraction. Its alkylated derivative S-alkylthiosulfonates are called Bunte salts. It is involved in the preparation of thioglycolic acid. Furthermore, it is used to dechlorinate tap water, for removing free bromine from solution and to check pH of bleach solutions with liquid indicators. Other used include tanning of leather, as an active component in hand warmers, in chemical heating pads and as an anionic surfactant to aid in dispersion of pharmaceutical preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium thiosulfate pentahydrate is used as an analytical reagent in laboratories. It is used in photographic processing and in gold extraction. Its alkylated derivative S-alkylthiosulfonates are called Bunte salts. It is involved in the preparation of thioglycolic acid. Furthermore, it is used to dechlorinate tap water, for removing free bromine from solution and to check pH of bleach solutions with liquid indicators. Other used include tanning of leather, as an active component in hand warmers, in chemical heating pads and as an anionic surfactant to aid in dispersion of pharmaceutical preparations.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Incompatible with sodium nitrate, acids, strong oxidizing agents, iodides, silver salts, mercury salts and lead.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text