Thermo Scientific Chemicals

2,4-Difluorobenzoyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: A17920.14
25 g, Each
Thermo Scientific Chemicals

2,4-Difluorobenzoyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: A17920.14
25 g, Each
Quantity
Catalog number: A17920.14
also known as A17920-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
72482-64-5
IUPAC Name
2,4-difluorobenzoyl chloride
Molecular Formula
C7H3ClF2O
InChI Key
JSWRVDNTKPAJLB-UHFFFAOYSA-N
SMILES
FC1=CC=C(C(Cl)=O)C(F)=C1
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.5120-1.5170 @ 20?C
Form
Liquid
Assay (GC)
≥96.0%

Description

2,4-Difluorobenzoyl chloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Difluorobenzoyl chloride is used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from water/ moisture, oxidizing agents, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text