Lithium acetate dihydrate is used as a buffer component in the modified ninhydrin reagent for amino analysis by ion-exchange chromatography. It is used as a catalyst in organic reactions, alkyd resin and acrylic polymer production and as an anticorrosion agent in molding polyphenylene sulfide resins as well as in the separation of saturated and unsaturated fatty acids. In the pharmaceutical industry, it is utilized for the preparation of diuretics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Lithium acetate dihydrate is used as a buffer component in the modified ninhydrin reagent for amino analysis by ion-exchange chromatography. It is used as a catalyst in organic reactions, alkyd resin and acrylic polymer production and as an anticorrosion agent in molding polyphenylene sulfide resins as well as in the separation of saturated and unsaturated fatty acids. In the pharmaceutical industry, it is utilized for the preparation of diuretics.
Solubility
Soluble in water and ethanol.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
- Schroder, F.; Winkler, B.; Bauer, J. D.; Haussuhl, E.; Escoto, B. R.; Lopez, F. T.; Borja, M. A.; Richter, C.; Ferner, J. Evidence for low-temperature isotopic ordering in methyl-monodeuterated lithium acetate dihydrate. J. Chem. Thermodyn. 2012, 54, 373-376.
- Winkler, B.; Haussuhl, E.; Bauer, J. D.; Schroder, F.; Refson, K.; Milman, V.; Hennion, B.; Bossak, A. Krisch, M. Influence of deuteration on lithium acetate dihydrate studied by inelastic X-ray scattering, density functional theory, thermal expansion, elastic and thermodynamic measurements. Dalton Trans. 2011, 40 (8), 1737-1742.