Thermo Scientific Chemicals

1-Heptylamine, 98+%, Thermo Scientific Chemicals

Catalog number: A17922.14
25 g, Each
Thermo Scientific Chemicals

1-Heptylamine, 98+%, Thermo Scientific Chemicals

Catalog number: A17922.14
25 g, Each
Quantity
Catalog number: A17922.14
also known as A17922-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-68-2
IUPAC Name
heptan-1-amine
Molecular Formula
C7H17N
InChI Key
WJYIASZWHGOTOU-UHFFFAOYSA-N
SMILES
CCCCCCCN
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Refractive Index
1.4230-1.4270 @ 20?C

Description

1-Heptylamine has been used as an internal standard in the determination of biogenic monoamines and biogenic diamines by the fluorescence-HPLC method. It has also been used to study the effect of hydration on the gaseous structure of protonated heptylamine by infrared photo dissociation (IRPD) spectroscopy and computational chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Heptylamine has been used as an internal standard in the determination of biogenic monoamines and biogenic diamines by the fluorescence-HPLC method. It has also been used to study the effect of hydration on the gaseous structure of protonated heptylamine by infrared photo dissociation (IRPD) spectroscopy and computational chemistry.

Solubility
Soluble in water 6791 mg/L @ 25°C.

Notes
Air sensitive. Store away from oxidizing agents, air, acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text