Triethylamine hydrobromide, 98%, Thermo Scientific Chemicals
Triethylamine hydrobromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethylamine hydrobromide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number A17926.36
also known as A17926-36
Price (USD)/ Each
148.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
148.00
Add to cart
Triethylamine hydrobromide, 98%, Thermo Scientific Chemicals
Catalog numberA17926.36
Price (USD)/ Each
148.00
-
Add to cart
Chemical Identifiers
CAS636-70-4
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Bromide)> 97.5%
Formcrystalline powder
Loss on Drying< 1.0% (105?C, constant weight)
Appearance (Color)White
Triethylamine hydrobromide is used to prepare dimethyl -alfa-diethylaminomaleate by reacting with dimethyl acetylenedicarbosylate. It is involved in the preparation of triphenylpropargylphosphonium bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylamine hydrobromide is used to prepare dimethyl -alfa-diethylaminomaleate by reacting with dimethyl acetylenedicarbosylate. It is involved in the preparation of triphenylpropargylphosphonium bromide.

Solubility
Soluble in water, alcohol and chloroform.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and bases.
RUO – Research Use Only

General References:

  1. Liu, D.; Li, D.; Du, D.; Zhao, X.; Qin, A.; Li, X.; He, C. Antifouling PVDF membrane with hydrophilic surface of terry pile-like structure. J. Membr. Sci. 2015, 493, 243-251.
  2. Qiu, M.; Zhao, X. Z.; Liu, D. P.; He, C. J. pH sensitive amphiphilic conetworks based on end-group cross-linking of polydimethylsiloxane pentablock copolymer and polymethylhydrosiloxane. RSC Adv. 2015, 5 (23), 17851-17861.