Thermo Scientific Chemicals

Squalane, 98%, Thermo Scientific Chemicals

Catalog number: A17931.22
100 g, Each
Thermo Scientific Chemicals

Squalane, 98%, Thermo Scientific Chemicals

Catalog number: A17931.22
100 g, Each
Quantity
Catalog number: A17931.22
also known as A17931-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
111-01-3
IUPAC Name
2,6,10,15,19,23-hexamethyltetracosane
Molecular Formula
C30H62
InChI Key
PRAKJMSDJKAYCZ-UHFFFAOYNA-N
SMILES
CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.4500-1.4540 @ 20?C

Description

Squalane influence on the vital pathway that transforms glucose into cholesterol may be pivotal. It regulates or controls the rate of synthesis of the enzyme HMG Co-A reductase, and may contribute to effective treatments for cancer and heart disease. It plays an important role as a terminator in the biomembrane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text