Thermo Scientific Chemicals

Diphenyliodonium hexafluorophosphate, 98%, Thermo Scientific Chemicals

Catalog number: A17934.18
50 g, Each
Thermo Scientific Chemicals

Diphenyliodonium hexafluorophosphate, 98%, Thermo Scientific Chemicals

Catalog number: A17934.18
50 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A17934.18
also known as A17934-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
58109-40-3
Appearance (Color)
White to pale cream
Melting Point (clear melt)
138-145?C
Form
Crystals or powder or crystalline powder
Elemental Analysis
Carbon: 32.98-34.67% (Theory: 33.83%)
Identification (FTIR)
Conforms

Description

Diphenyliodonium hexafluorophosphate is used as cationic photoinitiator and photoacid generator. It acts as a catalyst for the photochemical polymerization of various monomers. Further, it used as arylating reagent as well as an oxidant in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyliodonium hexafluorophosphate is used as cationic photoinitiator and photoacid generator. It acts as a catalyst for the photochemical polymerization of various monomers. Further, it used as arylating reagent as well as an oxidant in organic synthesis.

Solubility
Soluble in tetrahydrofuran, ethyl acetate, methanol, acetonitrile, dimethylformamide, dimethyl carbonate. Sparingly soluble in chloroform and dichloromethane. Insoluble in pentanes, hexanes, toluene and diethyl ether.

Notes
Light sensitive and hygroscopic. Incompatible with strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text