1x1 image pixel for data collection
Diphenyliodonium hexafluorophosphate, 98%
Diphenyliodonium hexafluorophosphate, 98%
Diphenyliodonium hexafluorophosphate, 98%
Diphenyliodonium hexafluorophosphate, 98%
Thermo Scientific Chemicals

Diphenyliodonium hexafluorophosphate, 98%

CAS: 58109-40-3 | C12H10I | 281.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A17934.30
also known as A17934-30
Price (USD)
1,401.00
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
1,401.00
Each
Add to cart
Chemical Identifiers
CAS58109-40-3
IUPAC Namediphenyliodanium
Molecular FormulaC12H10I
InChI KeyOZLBDYMWFAHSOQ-UHFFFAOYSA-N
SMILES[I+](C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)136-145°C
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Elemental AnalysisCarbon: 32.98-34.67% (Theory: 33.83%)
Identification (FTIR)Conforms
Diphenyliodonium hexafluorophosphate is used as cationic photoinitiator and photoacid generator. It acts as a catalyst for the photochemical polymerization of various monomers. Further, it used as arylating reagent as well as an oxidant in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyliodonium hexafluorophosphate is used as cationic photoinitiator and photoacid generator. It acts as a catalyst for the photochemical polymerization of various monomers. Further, it used as arylating reagent as well as an oxidant in organic synthesis.

Solubility
Soluble in tetrahydrofuran, ethyl acetate, methanol, acetonitrile, dimethylformamide, dimethyl carbonate. Sparingly soluble in chloroform and dichloromethane. Insoluble in pentanes, hexanes, toluene and diethyl ether.

Notes
Light sensitive and hygroscopic. Incompatible with strong acids.
RUO – Research Use Only

General References:

  1. Cahard, E.; Male, H. P. J.; Tissot, M.; Gaunt, M. J. Enantioselective and Regiodivergent Copper-Catalyzed Electrophilic Arylation of Allylic Amides with Diaryliodonium Salts. J. Am. Chem. Soc. 2015, 137 (25), 7986-7989.
  2. He, C.; Stoykovich, M. P. Photopatterning of Cross-Linkable Epoxide-Functionalized Block Copolymers and Dual-Tone Nanostructure Development for Fabrication Across the Nano-and Microscales. Small 2015, 11 (20), 2407-2416.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link