4-Fluoro-1,2-dimethoxybenzene, 98%, Thermo Scientific Chemicals
4-Fluoro-1,2-dimethoxybenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Fluoro-1,2-dimethoxybenzene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A17936.14
also known as A17936-14
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
Add to cart
4-Fluoro-1,2-dimethoxybenzene, 98%, Thermo Scientific Chemicals
Catalog numberA17936.14
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS398-62-9
IUPAC Name4-fluoro-1,2-dimethoxybenzene
Molecular FormulaC8H9FO2
InChI KeyDAGKHJDZYJFWSO-UHFFFAOYSA-N
SMILESCOC1=CC=C(F)C=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5080-1.5120 @ 20?C
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to pale yellow
4-Fluoro-1,2-dimethoxybenzene is a raw material used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-1,2-dimethoxybenzene is a raw material used in organic synthesis.

Solubility
Soluble in water 0.079 g/L 25°C.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Shizuka Saito; Jun Kawabata. Effects of electron-withdrawing substituents on DPPH radical scavenging reactions of protocatechuic acid and its analogues in alcoholic solvents. Tetrahedron. 2005, 61, (34), 8101-8108.
  2. Momoe Adachi; Koichi Tanaka and Koji Sekai. Aromatic Compounds as Redox Shuttle Additives for 4 V Class Secondary Lithium Batteries. J. Electrochem. Soc. 1999, 146, (4), 1256-1261.