Malonamide, 98%, Thermo Scientific Chemicals
Malonamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Malonamide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
100 g
500 g
Catalog number A17939.0E
also known as A17939-0E
Price (USD)/ Each
1,019.00
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
1,019.00
Add to cart
Malonamide, 98%, Thermo Scientific Chemicals
Catalog numberA17939.0E
Price (USD)/ Each
1,019.00
-
Add to cart
Chemical Identifiers
CAS108-13-4
IUPAC Namepropanediamide
Molecular FormulaC3H6N2O2
InChI KeyWRIRWRKPLXCTFD-UHFFFAOYSA-N
SMILESNC(=O)CC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Elemental Analysis(Nitrogen) : 26.75 - 28.12 %
Appearance (Color)White to very pale cream
Elemental Analysis(Carbon) : 34.42 - 36.17 %
FormCrystals or crystalline powder
Purity (DSC)>97.5%
View more
Malonamide is a reagent for fluorimetric determination of reducing carbohydrates. The malonamide-based ionic liquid extractant was used in the extraction of europium(iii) and other trivalent rare earth elements from nitric acid medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Malonamide is a reagent for fluorimetric determination of reducing carbohydrates. The malonamide-based ionic liquid extractant was used in the extraction of europium(iii) and other trivalent rare earth elements from nitric acid medium.

Solubility
Soluble in HCl.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Carmen E. Lisowski and James E. Hutchison. Malonamide-Functionalized Gold Nanoparticles for Selective, Colorimetric Sensing of Trivalent Lanthanide Ions. Anal. Chem. 2009, 81, (24), 10246-10253.
  2. C. Erlinger; L. Belloni; Th. Zemb; and C. Madic. Attractive Interactions between Reverse Aggregates and Phase Separation in Concentrated Malonamide Extractant Solutions. Langmuir. 1999, 15, (7), 2290-2300.
  3. Reagent for fluorimetric determination of reducing carbohydrates: Anal. Chim. Acta, 108, 421 (1979).