Hydroxypropyl methacrylate is a functional monomer for the preparation of hot solid acrylic coatings, styrene-butadiene latex modifiers, acrylic modified polyurethane coating ,water-soluble plating coatings , adhesives ,textile finishing agent ,paper coating , photosensitive paint and polyurethane vinyl resin modified agent. Mainly used in the manufacture are active groups of hydroxyl acrylic resin. Used in reactive diluent and cross-linking agent in the UV curing system.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Hydroxypropyl methacrylate is a functional monomer for the preparation of hot solid acrylic coatings, styrene-butadiene latex modifiers, acrylic modified polyurethane coating ,water-soluble plating coatings , adhesives ,textile finishing agent ,paper coating , photosensitive paint and polyurethane vinyl resin modified agent. Mainly used in the manufacture are active groups of hydroxyl acrylic resin. Used in reactive diluent and cross-linking agent in the UV curing system.
Solubility
Soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
Elizabeth H. Leduc and S. J. Holt. Hydroxypropyl Methacrylate, A New Water-Miscible Embedding Medium For Electron Microscopy. JCB Home. 1965, 26, (1), 137-155.
Murakami T; Itoshima T; Hitomi K; Ohtsuka A; Jones AL. A monomeric methyl and hydroxypropyl methacrylate injection medium and its utility in casting blood capillaries and liver bile canaliculi for scanning electron microscopy.. Arch Histol Jpn. 1984, 47, (2), 223-237.