Thermo Scientific Chemicals

Dimethylphenylphosphine, 96%, Thermo Scientific Chemicals

Catalog number: A17975.03
1 g, Each
Thermo Scientific Chemicals

Dimethylphenylphosphine, 96%, Thermo Scientific Chemicals

Catalog number: A17975.03
1 g, Each
Quantity
Catalog number: A17975.03
also known as A17975-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
672-66-2
IUPAC Name
dimethyl(phenyl)phosphane
Molecular Formula
C8H11P
InChI Key
HASCQPSFPAKVEK-UHFFFAOYSA-N
SMILES
CP(C)C1=CC=CC=C1
Assay (GC)
≥95.0%
Refractive Index
1.5610-1.5660 @ 20°C
Form
Liquid
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms

Description

Dimethylphenylphosphine is used as a ligand in transition metal complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylphenylphosphine is used as a ligand in transition metal complexes.

Solubility
Soluble in organic solvents.

Notes
Air sensitive. Store away from oxidizing agents, air, acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text