Thermo Scientific Chemicals

4-Phenoxybenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A17998.14
25 g, Each
Thermo Scientific Chemicals

4-Phenoxybenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A17998.14
25 g, Each
Quantity
Catalog number: A17998.14
also known as A17998-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2215-77-2
IUPAC Name
4-phenoxybenzoic acid
Molecular Formula
C13H10O3
InChI Key
RYAQFHLUEMJOMF-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1
Assay (Silylated GC)
≥98.5%
Melting Point (clear melt)
159.0-165.0?C
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

4-Phenoxybenzoic acid can block DNA binding of the human papillomarvirus (HPV) E2 protein. It is employed as an intermediate of Sitafloxacin, which is a fluoroquinolone antibiotic that shows promise in the treatment of Buruli ulcer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenoxybenzoic acid can block DNA binding of the human papillomarvirus (HPV) E2 protein. It is employed as an intermediate of Sitafloxacin, which is a fluoroquinolone antibiotic that shows promise in the treatment of Buruli ulcer.

Solubility
Very faint turbidity in hot methanol.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text