Thermo Scientific Chemicals

m-Cresol Purple sodium salt, Thermo Scientific Chemicals

Catalog number: A18025.14
25 g, Each
Thermo Scientific Chemicals

m-Cresol Purple sodium salt, Thermo Scientific Chemicals

Catalog number: A18025.14
25 g, Each
Quantity
Catalog number: A18025.14
also known as A18025-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
62625-31-4
IUPAC Name
sodium 4-[3-(4-hydroxy-2-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-3-methylbenzen-1-olate
Molecular Formula
C21H17NaO5S
InChI Key
ALWHNCXHKCVHCK-UHFFFAOYNA-M
SMILES
[Na+].CC1=CC(O)=CC=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C([O-])C=C1C
Appearance (Color)
Dark orange to red-brown to brown to black
Form
Powder
Color Change
From pH 1.2 (red) to pH 2.8 (yellow); From pH 7.4 (yellow) to pH 9.0 (purple)
Solution Test
Clear, deep purple solution (1 mg/mL, 1N NH₄OH)

Description

m-Cresol purple sodium salt is used to detect changes in pH during titrations at which color changes occur at pH 7.6-9.2. Upon bromination, m-cresol purple sodium salt creates the dye bromocresol green (sc-206036) which is used as a tracking dye in DNA agarose gel electrophoresis. In addition, m-cresol purple sodium salt can be used to stain epithelial cells and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Cresol purple sodium salt is used to detect changes in pH during titrations at which color changes occur at pH 7.6-9.2. Upon bromination, m-cresol purple sodium salt creates the dye bromocresol green (sc-206036) which is used as a tracking dye in DNA agarose gel electrophoresis. In addition, m-cresol purple sodium salt can be used to stain epithelial cells and proteins.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text