Thermo Scientific Chemicals

(+/-)-Camphene, tech. (sum of camphene + fenchene), Thermo Scientific Chemicals

Catalog number: A18040.0B
1000 g, Each
Thermo Scientific Chemicals

(+/-)-Camphene, tech. (sum of camphene + fenchene), Thermo Scientific Chemicals

Catalog number: A18040.0B
1000 g, Each
Quantity
Catalog number: A18040.0B
also known as A18040-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79-92-5
IUPAC Name
2,2-dimethyl-3-methylidenebicyclo[2.2.1]heptane
Molecular Formula
C10H16
InChI Key
CRPUJAZIXJMDBK-UHFFFAOYNA-N
SMILES
CC1(C)C2CCC(C2)C1=C
Assay (HPLC)
≥78%
Appearance (Color)
Clear colorless or white to yellow
Form
Fused solid or liquid as melt

Description

Camphene is used in the preparation of fragrances and as a food additive for flavoring.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Camphene is used in the preparation of fragrances and as a food additive for flavoring.

Solubility
Insoluble in water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text