Thermo Scientific Chemicals

5-Bromovaleryl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A18046.06
5 g, Each
Thermo Scientific Chemicals

5-Bromovaleryl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A18046.06
5 g, Each
Quantity
Catalog number: A18046.06
also known as A18046-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4509-90-4
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.4895-1.4935 @ 20?C

Description

5-Bromovaleryl chloride was used in the preparation of aromatic azides and organic gelators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromovaleryl chloride was used in the preparation of aromatic azides and organic gelators.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from water/ moisture, oxidizing agents, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text