Thermo Scientific Chemicals

CHES, 99%

Catalog number: A18047.22
100 g, Each
Thermo Scientific Chemicals

CHES, 99%

Catalog number: A18047.22
100 g, Each
Quantity
Catalog number: A18047.22
also known as A18047-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
103-47-9
IUPAC Name
sodium 2-(cyclohexylamino)ethane-1-sulfonate
Molecular Formula
C8H16NNaO3S
InChI Key
TXWKJOVDGFRVOG-UHFFFAOYSA-M
SMILES
[Na+].[O-]S(=O)(=O)CCNC1CCCCC1
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤0.5%
Appearance (Color)
White

Description

CHES is zwitterionic buffer useful in pH range 8.6 to 10.0.CHES is widely utilized as a buffer which is used for investigations on pH-dependent processes in enzymology. It has been shown to have an unusually high affinity for the iodoacetate binding site of liver alcohol dehydrogenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
CHES is zwitterionic buffer useful in pH range 8.6 to 10.0.CHES is widely utilized as a buffer which is used for investigations on pH-dependent processes in enzymology. It has been shown to have an unusually high affinity for the iodoacetate binding site of liver alcohol dehydrogenase.

Solubility
Soluble in water.

Notes
Store in cool place. Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text